Learn More
Kimble™ KIMAX™ Flasks with Graduation Line Etched through Vertical Red Stripe Available on GSA/VA Contract for Federal Government customers only.
Calibrated “To Contain” within Class A tolerances
Manufacturer:  Kimble™ 2801325
 View more versions of this product
Catalog No. 10-248A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Calibrated To Contain
9
4.64 in.
±0.03mL
Borosilicate Glass
Glass
Yes
11.8cm
25mL