Learn More
DWK Life Sciences KIMBLE™ RAY-SORB™ Amber Class A Volumetric Flask
Calibrated “To Contain”
$919.00 - $2700.00
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
10-204-6
|
DWK Life Sciences
28016200 |
200 mL |
Case of 12 for $1,153.00
|
|
10-229
|
DWK Life Sciences
2801610 |
10 mL |
Case of 12 for $919.00
|
|
10-229A
|
DWK Life Sciences
2801625 |
25 mL |
Case of 12 for $1,047.00
|
|
10-229B
|
DWK Life Sciences
2801650 |
50 mL |
Case of 12 for $1,123.00
|
|
10-229D
![]() |
DWK Life Sciences
28016250 |
250 mL |
Case of 12 for $1,444.00
|
|
10-229E
![]() |
DWK Life Sciences
28016500 |
500 mL |
Case of 12 for $2,700.00
|
|
10-229C
|
DWK Life Sciences
28016100 |
100 mL |
Case of 12 for $1,119.00
|
|
10-229F
![]() |
DWK Life Sciences
280161000 |
1000 mL |
Case of 6 for $1,639.00
|
|
Description
- Protection from wavelengths below 500nm for materials sensitive to light
- Class A, RAY-SORB™ processed glass
- Marking spots
- PTFE, standard taper stopper with color-coded handle
- Stopper standard taper sizes: 9 (10, 25, 50mL); 13 (100); 16 (200, 250); 19 (500); 22 (1000)
- Tolerances specified on Class A Tolerances Chart
Specifications
Borosilicate Glass |