Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ KIMAX™ Flasks with Pennyhead standard taper Stopper
For volatile or hygroscopic liquids
Supplier: DWK Life Sciences 26600125
Description
- Narrow mouths
- Graduated scales
Compliance
Designed from ASTM™ E 1404, Type II, Class 2 requirements
Specifications
Glass | |
2.59 in. | |
Borosilicate Glass | |
25 mL | |
11.8 cm | |
Penny Stopper | |
22 |
Capacity scale | |
125 mL | |
50 to 125 mL | |
4.64 in. | |
Narrow | |
66 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction