Learn More
Kimble™ KIMAX™ Flasks with standard taper Pennyhead Stoppers Available on GSA/VA Contract for Federal Government customers only.
Calibrated “To Contain”
Manufacturer:  Kimble™ 2801425
 View more versions of this product
Catalog No. 10-209B
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Calibrated To Contain
9
11.8cm
25mL
Glass
4.64 in.
±0.03mL
Borosilicate Glass