Learn More
Kimble™ KIMAX™ Flasks with standard taper Pennyhead Stoppers Available on GSA/VA Contract for Federal Government customers only.
Calibrated “To Contain”
Manufacturer:  Kimble™ 28014200
 View more versions of this product
Catalog No. 10-209E
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Calibrated To Contain
16
22.5cm
200mL
Glass
8.85 in.
±0.1mL
Borosilicate Glass