Learn More
DWK Life Sciences KIMBLE™ RAY-SORB™ Amber Heavy Duty Wide Mouth Volumetric Flask, Glass Stopper
SDP
Protects contents from harmful UV light
$355.00 - $1084.00
Specifications
Closure Material Glass
Color Red
Material Borosilicate Glass
Mouth Style Wide
Bottom Shape Flat
View More Specs
Products 11
Catalog Number Mfr. No. Capacity (Metric) Closure Size Tolerance Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Tolerance Price Quantity & Availability  
13-756-700
SDP
View Documents
DWK Life Sciences
92822G5
5 mL 13 ±0.08 mL
Case of 6 for $468.00
Add to Cart
13-756-701
SDP
View Documents
DWK Life Sciences
92822G10
10 mL 13 ±0.08 mL
Case of 6 for $529.00
Add to Cart
13-756-706
SDP
View Documents
DWK Life Sciences
92822G200
200 mL 19 ±0.2 mL
Case of 6 for $874.00
Add to Cart
13-756-705
SDP
View Documents
DWK Life Sciences
92822G100
100 mL 16 ±0.1 mL
Case of 6 for $796.00
Edge
Add to Cart
13-756-704
SDP
View Documents
DWK Life Sciences
92822G50
50 mL 13 ±0.08 mL
Case of 6 for $589.00
Add to Cart
13-756-702
SDP
View Documents
DWK Life Sciences
92822G20
20 mL 13 ±0.08 mL
Case of 6 for $568.00
Add to Cart
13-756-703
SDP
View Documents
DWK Life Sciences
92822G25
25 mL 13 ±0.08 mL
Case of 6 for $568.00
Add to Cart
13-756-708
SDP
View Documents
DWK Life Sciences
92822G500
500 mL 19 ±0.2 mL
Case of 6 for $1,084.00
Add to Cart
13-756-707
SDP
View Documents
DWK Life Sciences
92822G250
250 mL 19 ±0.2 mL
Case of 6 for $911.00
Add to Cart
13-756-710
SDP
View Documents
DWK Life Sciences
92822G2000
2000 mL 27 ±0.5 mL
Case for $520.00
Add to Cart
13-756-709
SDP
View Documents
DWK Life Sciences
92822G1000
1000 mL 22 ±0.3 mL
Case for $355.00
Add to Cart
Description
Description
  • Wide mouth makes pipet access easy
  • Heavy, uniform walls provide superior strength, durability and safety
  • Large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Calibrated “To Contain”
  • Sizes from 5 to 250mL are calibrated to meet ASTM™ E288
Specifications
Specifications
Glass
Borosilicate Glass
Flat
Red
Wide
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.