Learn More
Kimble™ KIMAX™ Heavy-Duty RAY-SORB™ Glass Volumetric Class A Flasks with PTFE Stoppers
Stoppers are color coded for easy identification
Manufacturer:  Kimble™ 92822F1000
 View more versions of this product
Catalog No. 13-756-720
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Flat
PTFE
Red
32.8cm
±0.3mL
Borosilicate Glass
Yellow
22
12.9 in.
Wide
1000mL