Learn More
DWK Life Sciences KIMBLE™ RAY-SORB™ Amber Heavy Duty Wide Mouth Volumetric Flask, No Stopper
RAY-SORB processing protects flask contents from short-length lightwaves
$305.00 - $944.00
Specifications
Color | Red |
---|---|
Material | Borosilicate Glass |
Mouth Style | Wide |
Bottom Shape | Flat |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
13-757-833
|
DWK Life Sciences
92822N5 |
5 mL |
Case of 6 for $448.00
|
|
|||||
13-757-837
|
DWK Life Sciences
92822N50 |
50 mL |
Case of 6 for $511.50
|
|
|||||
13-757-838
|
DWK Life Sciences
92822N100 |
100 mL |
Case of 6 for $698.00
|
|
|||||
13-757-834
|
DWK Life Sciences
92822N10 |
10 mL |
Case of 6 for $458.50
|
|
|||||
13-757-835
|
DWK Life Sciences
92822N20 |
20 mL |
Case of 6 for $473.00
|
|
|||||
13-757-840
|
DWK Life Sciences
92822N250 |
250 mL |
Case of 6 for $807.00
|
|
|||||
13-757-839
|
DWK Life Sciences
92822N200 |
200 mL |
Case of 6 for $774.00
|
|
|||||
13-757-836
|
DWK Life Sciences
92822N25 |
25 mL |
Case of 6 for $473.00
|
|
|||||
13-757-841
|
DWK Life Sciences
92822N500 |
500 mL |
Case of 6 for $944.00
|
|
|||||
13-757-842
|
DWK Life Sciences
92822N1000 |
1000 mL |
Case for $305.00
|
|
|||||
13-757-843
|
DWK Life Sciences
92822N2000 |
2000 mL |
Case for $461.50
|
|
|||||
Description
- Wide mouth makes pipet access easy
- Heavy, uniform walls provide superior strength, durability and safety
- Large, permanent, easy-to-read markings
- Flat interior bottoms for stirring with standard stir bars
- Graduated to Class A volumetric tolerances
- Calibrated “To Contain”
- Sizes from 5 to 250mL are calibrated to meet ASTM™ E288
Specifications
Red | |
Borosilicate Glass | |
Wide | |
Flat |