Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Kimble™ KIMAX™ Nonvacuum Glass Desiccators

Glass desiccators made for durability and efficiency in daily use

Manufacturer:  Kimble™ 21050200

 View more versions of this product

Catalog No. 08-595-5A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: Cover

Requires : Desiccant plate

Description

Description

  • Jars shaped to hold sample plate (not supplied) above desiccant
  • Bottoms sloped so desiccant can be removed easily
  • Manufactured from 33 expansion borosilicate glass

Specifications

Specifications

3.8L
20cm
31cm
Borosilicate Glass
Knob top
Nonvacuum Desiccator
USP Type I, ASTM E438, Type I, Class A
19cm
Cover
Medium
20cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit