Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ KIMAX™ Soxhlet Apparatus with Allihn Condenser, Complete unit

Designed from Fed. Spec. NNN-F-240

Manufacturer:  DWK Life Sciences 2400550

 View more versions of this product

Catalog No. 09-551C


Add to cart

Description

Description

  • Complete assembly includes 09-551-10 Series Allihn condenser, 09-553 Series Soxhlet extraction tube with standard taper 24/40 joint at bottom, 09-552 Series flask
  • All connected by interchangeable ground joints
  • Hose connections on condenser accept 0.31 in. I.D. (8mm) flexible tubing

Specifications

Specifications

16.9 oz.
16.9 oz., 500mL
72cm
1.96 in.
55/50
Extraction Apparatus Assembly
500mL
28.34 in.
Allihn condenser, soxhlet extraction tube
5cm
43 dia. x 123mm H paper thimble
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit