Learn More
DWK Life Sciences Kimble™ KIMAX™ Squibb Funnels with standard taper Stopcock and Stopper Available on GSA/VA Contract for Federal Government customers only.
Lower stems made of large-diameter tubing for liquid to drain out completely when stopcock is closed
Manufacturer:  DWK Life Sciences 29048125
 View more versions of this product
Catalog No. 10-435-5B
Add to cart
Description
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ASTM E1096, Type IV
Squibb separatory funnel
2; Glass
22
Borosilicate Glass
60mm
2 Pack
Glass
125 mL