Learn More
Kimble™ KIMAX™ Standard Wall Borosilicate Glass Tubing
Low alkali content borosilicate glass tubing for general laboratory use
Manufacturer:  Kimble™ 80200 10
 View more versions of this product
Catalog No. 11-368H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
10mm
Borosilicate Glass
0.39 in.
1219mm
1mm
2291°F
48 in.
Type I Class A, ASTM E438-92
For general laboratory use
Standard Wall Tubing
±0.1mm
1255°C