Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Accuform™ SSR™ Standard Glass Vials

Vials provide dependable storage, retrevial and analysis of both liquid and powder samples

Manufacturer:  DWK Life Sciences 606904

 View more versions of this product

Catalog No. 06-100-258

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Borosilicate glass construction for chemical resistance
  • Large opening tops for easy access of contents
  • Less than 10µL of dead space
  • Smooth, conical interior surfaces facilitate complete sample recovery
  • Smooth bottom allows for 2D barcoding
  • Black phenolic caps work with media bottles and screw-thread vials
  • Offered in two different styles—stand and straight sided

Specifications

Specifications

Open screw top
24-100
Borosilicate Glass
16 mL
70mm
Conical
4.32 dr.
USP Type I, ASTM E438, Type I, Class A
21 mm
SSR™ Straight-sided Vial
Ideal for high throughput screening
70mm (2.75 in.)
24-100
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit