Learn More
Kimble™ Serological Disposable Pipet
Sterile, color coded
Manufacturer:  Kimble™ 7210221100
 View more versions of this product
Catalog No. 13-678-251
  Check Availability
Add to cart
Specifications
Description
  • Borosilicate glass
  • Plugged
  • Calibrated
  • Non-Pyrogenic
Description & Specifications
Specifications
0.2mL
2mL
Borosilicate Glass
ASTM E714
Yes
±0.06mL
0.06 oz.
Yes
White
Serological Pipet
0.01mL
0.06 oz., 2mL