Learn More
DWK Life Sciences Kimble™ Kontes™ 20mm Flip-Off Button-Top Aluminum Seals, Color Coded
Two-piece, tamper-evident flip-off feature protects target area
Manufacturer:  DWK Life Sciences 73845B20
 View more versions of this product
Catalog No. 03-340-337
  Check Availability
Add to cart
Description
Description
  • Button top is easily removed with the flip of a thumb
  • For use with 20mm serum finished bottles and vials
  • Designed to secure septa and stoppers to bottles and vials
  • Available with blue, red, green and white plastic buttons to help separate and identify samples
Specifications
Specifications
Flip-up Button-top
Aluminum
1000 Case
Aluminum
Yes
Blue
Seal
20mm