Learn More
DWK Life Sciences Kimble™ Kontes™ Center Tear-Out Aluminum Seals, Color Coded
Designed to secure septa and stoppers to bottles and vials
Manufacturer:  DWK Life Sciences 73820B20
 View more versions of this product
Catalog No. 03-340-315
Add to cart
Description
Description
  • Tamper-evident center tear-out feature protects target area
  • For use with 13 and 20mm serum finished bottles and vials
  • Available in blue, red, and green color to help separate and identify samples
Specifications
Specifications
Aluminum
1000 Case
Tear-out One Piece Seal
Tear-out
20mm
Blue