Learn More
DWK Life Sciences Kimble™ Kontes™ Flat Bottom Heavy-Wall Boiling Flasks
Single neck with a standard tapered outer joint
Supplier: DWK Life Sciences 6015000829
Description
- Heavy wall reduces breakage.
Specifications
6.3 in. | |
Borosilicate Glass | |
29/42 | |
160 mm |
2000 mL | |
1 | |
Flat |