Learn More
DWK Life Sciences Kimble™ Kontes™ Flex-Column Economy Columns Accessory: Polypropylene Reservoirs
For use with Kimble-Chase Kontes Flex-Column Economy Columns
$293.00 - $564.00
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
K420405-0100
|
DWK Life Sciences
4204050100 |
0.7, 1 and 1.5cm I.D. FlexColumn™ economy column for initial column packing and as a small Buffer reservoir |
Case for $564.00
|
|
|||||
K420405-0700
|
DWK Life Sciences
4204050700 |
2.5cm I.D. FlexColumn™ economy column for initial column packing and as a small Buffer reservoir |
Case for $293.00
|
|
|||||
Description
For use with Kimble-Chase Kontes Flex-Column Economy Columns
- Available in two sizes: 100mL and 700mL
- 100mL: fits 0.7, 1.0, and 1.5cm diameter Flex-Column columns
- 700mL: fits 2.5cm columns
- For Flex-Column Economy Columns, see cat. no. K420400-1015
Specifications
Packing Reservoir | |
Polypropylene |