Learn More
DWK Life Sciences Kimble™ Kontes™ Flip-Off Button-Top Aluminum Seals, Color Coded
Two-piece, tamper-evident flip-off button top protects target area and allows easy access
Manufacturer:  DWK Life Sciences 73843D20
 View more versions of this product
Catalog No. 03-340-331
  Check Availability
Add to cart
Description
Description
  • For use with 13 and 20mm serum finished bottles and vials
  • Designed to secure septa and stoppers to bottles and vials
  • Allows easy access with a flip of the thumb
  • Available with blue, red, green and white plastic buttons to help separate and identify samples
Specifications
Specifications
Flip-up Button-top
Aluminum
1000 Case
Aluminum
Yes
Green
Seal
20mm