Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Flip-Off Button-Top Aluminum Seals, Color Coded
Two-piece, tamper-evident flip-off button top protects target area and allows easy access
Supplier: DWK Life Sciences W224179
This item is not returnable.
View return policy
Description
- For use with 13 and 20 mm serum finished bottles and vials
- Designed to secure septa and stoppers to bottles and vials
- Allows easy access with a flip of the thumb
- Available with blue, red, green, and white plastic buttons to help separate and identify samples
Specifications
Blue | |
Autoclavable | |
Flip-up Button-top |
Aluminum | |
1000 Cs. | |
13 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title