Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ Flip-Off Button-Top Aluminum Seals, Color Coded
SDP

Catalog No. 01257643 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Closure Size:
13 mm
20 mm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Size
01-257-643 Blue 20 mm
03-340-324 White 13 mm
03-340-328 White 20 mm
03-340-327 Green 13 mm
03-340-331 Green 20 mm
01-257-642 Blue 13 mm
03-340-326 Red 13 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 01-257-643 Supplier DWK Life Sciences Supplier No. W224180
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Two-piece, tamper-evident flip-off button top protects target area and allows easy access

  • For use with 13 and 20 mm serum finished bottles and vials
  • Designed to secure septa and stoppers to bottles and vials
  • Allows easy access with a flip of the thumb
  • Available with blue, red, green, and white plastic buttons to help separate and identify samples
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Aluminum
Quantity 1000 Cs.
Closure Size 20 mm
Autoclavable Autoclavable
Product Type Flip-up Button-top Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.