Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Flip-Off Button-Top Aluminum Seals, Color Coded
Two-piece, tamper-evident flip-off button top protects target area and allows easy access
$218.03 - $490.00
Specifications
Closure Material | Aluminum |
---|---|
Autoclavable | Autoclavable |
Closure Type | Flip-up Button-top |
Quantity | 1000 Case |
Catalog Number | Mfr. No. | Color | Thread Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Thread Size | Price | Quantity & Availability | ||||
03-340-324
|
DWK Life Sciences
73843A13 |
White | 13 mm |
Case of 1000 for $334.00
|
|
03-340-328
|
DWK Life Sciences
73843A20 |
White | 20 mm |
Case of 1000 for $490.00
|
|
03-340-326
|
DWK Life Sciences
73843C13 |
Red | 13 mm |
Case of 1000 for $334.00
|
|
03-340-327
|
DWK Life Sciences
73843D13 |
Green | 13 mm |
Case of 1000 for $334.00
|
|
03-340-331
|
DWK Life Sciences
73843D20 |
Green | 20 mm |
Case of 1000 for $490.00
|
|
01-257-642
|
DWK Life Sciences
W224179 |
Blue | 13 mm |
Case of 1000 for $218.03
|
|
01-257-643
|
DWK Life Sciences
W224180 |
Blue | 20 mm |
Case of 1000 for $321.70
|
|
03-340-330
|
DWK Life Sciences
73843C20 |
Red | 20 mm | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- For use with 13 and 20 mm serum finished bottles and vials
- Designed to secure septa and stoppers to bottles and vials
- Allows easy access with a flip of the thumb
- Available with blue, red, green, and white plastic buttons to help separate and identify samples
Specifications
Aluminum | |
Autoclavable | |
Flip-up Button-top | |
1000 Case |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title