Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ Flip-Off Button-Top Aluminum Seals, Color Coded

Two-piece, tamper-evident flip-off button top protects target area and allows easy access

Manufacturer:  DWK Life Sciences 73843D13

 View more versions of this product

Catalog No. 03-340-327


  Check Availability
Add to cart

Description

Description

  • For use with 13 and 20mm serum finished bottles and vials
  • Designed to secure septa and stoppers to bottles and vials
  • Allows easy access with a flip of the thumb
  • Available with blue, red, green and white plastic buttons to help separate and identify samples

Specifications

Specifications

Flip-up Button-top
Aluminum
1000 Case
Aluminum
Yes
Green
Seal
13mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit