Kimble™ Kontes™ Flip, Tear-Off Button Top Aluminum Seals, Color Coded
Designed to secure septa and stoppers to bottles and vials
Manufacturer: Kimble™ 73844A13
 View more versions of this product
Catalog No. 03-340-332
  Check Availability
Add to Cart
    Specifications
    Closure Material Aluminum
    Closure Type Flip-up Button-top
    Color White
    Quantity 1000 Case
    Type Seal
    Material Closure Aluminum
    Autoclavable Yes
    Thread Size 13mm
  • Description & Specifications
Related Products
Product Content Feedback