Learn More
DWK Life Sciences Kimble™ Kontes™ Flip, Tear-Off Button Top Aluminum Seals, Color Coded
Designed to secure septa and stoppers to bottles and vials
Manufacturer:  DWK Life Sciences 73844B13
 View more versions of this product
Catalog No. 03-340-333
  Check Availability
Add to cart
Description
Description
  • Tamper-evident flip, tear-off button top protects target area and allows easy removal of the aluminum seal
  • For use with 13mm serum finished bottles and vials
  • Available with blue, red, and white plastic buttons to help separate and identify samples
Specifications
Specifications
Flip-up Button-top
Aluminum
1000 Case
Aluminum
Yes
Blue
Seal
13mm