Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Heavy Wall Filtering Flask
Provides mechanical strength under vacuum application
Supplier: DWK Life Sciences 6175000003
Description
- Outer joint: 24/40
- Serrated hose connection: No. 2
Specifications
3.34 in. | |
250 mL | |
7.48 in. | |
85 mm |
0.93 cm | |
Borosilicate Glass | |
19 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title