Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Lids for Four Neck Reaction Flask
For use with cylindrical and spherical flasks having same flange I.D.
Supplier: DWK Life Sciences 6130004524
Description
- Flat ground
- Flange I.D. is 4 in. (10.16cm)
- Side standard taperjoint is 24/40
- Thermometer standard taperjoint is 10/30
- Uses clamp
Specifications
10.16 cm | |
Cylindrical and spherical flasks having the same I.D. | |
Cap |
Borosilicate Glass | |
4 | |
45/50 Center, 24/40 side |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title