Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ Kontes™ Micro-Vial Graduated Screw Thread Vials

With open-top closure and PTFE-faced septum

Manufacturer:  Kimble™ 607052

 View more versions of this product

Catalog No. 06-100-1B


  Check Availability
Add to cart

Specifications

Description

  • Accuform™
  • Borosilicate glass
  • Assembled

Description & Specifications

Specifications

Micro Vial
Borosilicate Glass
Open Top Screw Cap
12 Case
Yes
Yes
20-400
2mL
Amber
40mm
21mm
1.57 in.
Tapered
2.0mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit