Learn More
DWK Life Sciences Kimble Kontes NMR Pipets
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
$144.00
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
K897085-0009
![]() |
DWK Life Sciences
8970850009 |
NMR Pipet |
Case of 100 for $144.00
|
|
K897085-0007
|
DWK Life Sciences
8970850007 |
Extended Tip Pipet |
Case of 100 for $144.00
|
|
Description
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
- Available with 7 and 9 in. (17.7 and 23cm) tips for use with 7 and 9 in. long NMR tubes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Borosilicate Glass | |
100/Cs. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.