Learn More
DWK Life Sciences Kimble™ Kontes™ Polypropylene Closure with Ultrasonically Welded PTFE Liner
For use with screw-thread culture and centrifuge tubes
Supplier: DWK Life Sciences 7380828400
Description
- Provides superior closure system capable of maintaining its liner through multiple autoclavable operations
- PTFE-coated surface of exposed liner is chemical-resistant and compatible with sensitive, corrosive and volatile solvents
Specifications
28-400 | |
Screw Cap | |
Polypropylene | |
PTFE |
PTFE, Rubber | |
Autoclavable | |
For repeated autoclaving, to provide superior closure system | |
EPA Water Analysis Vials, Screw Thread Culture and Centrifuge Tubes |