Kimble™ Kontes™ PTFE Plug 14/20 Joints Micro Separatory Funnels with Stopper

With 2mm PTFE stopcock

Manufacturer: Kimble™ 2982800025

 View more versions of this product

Catalog No. 22-296626

  Check Availability
Add to Cart

    Specifications

    Product Type Micro Separatory Funnel
    Quantity 1 Case
    Capacity (Metric) 25mL
    Height (Metric) 171mm
    Capacity (English) 0.84 oz.
    Joint Number 14/20
    Material Borosilicate Glass
    Stopcock Number 2
    Height (English) 6.7 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit