Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Reaction Cylindrical Flask with Jacket
With flat ground flange
Supplier: DWK Life Sciences 6121000500
Description
- 3/16 in. wall construction
Specifications
10.16 cm | |
Borosilicate Glass | |
10 cm | |
Reaction cylindrical flask with jacket |
500 mL | |
3.93 in. | |
Flat ground flange and a 0.18 in. wall construction | |
Cylindrical |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title