Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ Round Bottom Heavy Wall Boiling Flasks
SDP

Single neck flask with a standard outer joint

Supplier:  DWK Life Sciences 6010000129

 View more versions of this product

Catalog No. K601000-0129


Add to Cart

Description

Description

  • Heavy wall reduces breakage.

Specifications

Specifications

1.8 in.
Borosilicate Glass
1
Round
50 mL
Single
29/42
48 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit