Learn More
DWK Life Sciences™ Kimble™ Open-Top Aluminum Seals, Color Coded
Designed to secure septa and stoppers to bottles and vials
Manufacturer:  DWK Life Sciences™ 73822C20
 View more versions of this product
Catalog No. 03-340-310
Add to cart
Description
Description
  • For use with 13 and 20mm serum finished bottles and vials
  • Available in blue, red, green and natural aluminum color to help separate and identify samples
Specifications
Specifications
Aluminum
20mm
Red
1000 Case
Open Top One Piece Seal