Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Open-Top Aluminum Seals, Color Coded Available on GSA/VA Contract for Federal Government customers only.

Designed to secure septa and stoppers to bottles and vials

Manufacturer:  DWK Life Sciences 73822C13

 View more versions of this product

Catalog No. 03-340-307


Add to cart

Description

Description

  • For use with 13 and 20mm serum finished bottles and vials
  • Available in blue, red, green and natural aluminum color to help separate and identify samples

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Aluminum
13mm
Red
1000 Case
Open Top One Piece Seal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit