Kimble™ Open-Top PolypropyleneScrew Thread Closures with Red PTFE-Faced Silicone Septa
Autoclavable and used for multiple injections
Manufacturer: Kimble™ 73813BK13425
 View more versions of this product
Catalog No. 03-341-613
  Check Availability
Add to Cart
    Specifications
    Color Black
    Pre-slit Yes
    Material Polypropylene
    GPI Thread 13-425
    Type Open Top Screw Cap
    Insert Type Pre-Slit
    Material Closure Polypropylene
    Autoclavable Yes
    Thread Size 13-425
  • Description & Specifications
Related Products
Product Content Feedback