Learn More
DWK Life Sciences Kimble™ Prelabeled Glass Blood Typing Mark-M-Tubes™
Made of borosilicate glass
Manufacturer:  DWK Life Sciences 60B12BZ1
 View more versions of this product
Catalog No. 22-285751
Add to cart
Description
Description
Fused labels make the written information permanent and easy to read. Tubes are prelabeled with the blood type, and a color-coded band with an area for patient identification. The marking area is placed to allow an unobstructed view of cell suspension.
Order Info
Available in two tube sizes: 10 x 75mm and 12 x 75mm.
Specifications
Specifications
ASTM E438
0.47 in.
A1 Testing
75mm
Borosilicate Glass
Yellow
12mm
2.9 in.
1000 Case
Blood Typing Tube
Documents
Accessories