Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ Clear Boston Rounds with Taperseal Caps

Narrow-mouth design provides more controlled pouring than wide-mouth.

Manufacturer:  Kimble™ 5110120V25

 View more versions of this product

Catalog No. 13-756-914


  Check Availability
Add to cart

Specifications

Description

  • High quality, economical narrow mouth bottles are manufactured from clear Type III soda-lime glass to ensure easy viewing of the contents
  • Supplied with caps

Description & Specifications

Specifications

1 oz., 30mL
1 oz.
Taperseal
1.22 in.
Glass
Boston Round Bottles
20-400
79mm
30mL
Phenolic
Clear
3.1 in.
Convenience Pack
Black
31mm
Screw
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit