Learn More
DWK Life Sciences Kimble™ Color Comparison APHA Standard Tubes
SDP
High-quality glass test tubes that meet Federal and American Public Health Association (APHA) standards
$331.00 - $427.00
Specifications
Material Borosilicate Glass
Certifications/Compliance NNN-T-175, USP, ASTM E 438, APHA
For Use With (Application) Used to fill with standard solution colors for visual color comparison with similar tubes containing solution sample
Includes White ceramic enamel rings and legend
Scale Type Graduated Ring
View More Specs
Products 2
Catalog Number Mfr. No. Graduations Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Graduations Length (Metric) Price Quantity & Availability  
22-043721
SDP
View Documents
DWK Life Sciences
4531550
50 mL 300 mm
Case of 6 for $331.00
Edge
Add to Cart
10-310-189B
SDP
View Documents
DWK Life Sciences
45315100
100 mL 375 mm
Case of 6 for $427.00
Add to Cart
Description
Description
High-quality glass test tubes that meet Federal and American Public Health Association (APHA) standards
  • “Shadowless Bottom” feature allows for viewing with undistorted light
  • Taller and more narrow with a longer scale length than earlier models
  • All graduation lines and legends are durable white ceramic enamel
  • Rings coincide with specific volumes of liquid
  • Comes in sets of six unmatched tubes in 50mL and 100mL sizes
  • Glass, nonsterile, autoclavable
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.
Compliance
Designed from Federal Specification NNN-T-175, Type 1, Class 2 requirements. The 50mL size is specifically mentioned by APHA in Standard Methods for the Examination of Water and Wastewater.
Specifications
Specifications
Borosilicate Glass
Used to fill with standard solution colors for visual color comparison with similar tubes containing solution sample
Graduated Ring
NNN-T-175, USP, ASTM E 438, APHA
White ceramic enamel rings and legend
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.