Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Disposable Drum Sampler

Borosilicate glass, open-end drum thieves break easily for quick disposal. Available in 150mL, 25mL and 75mL capacity.

Manufacturer:  DWK Life Sciences 7600025

 View more versions of this product

Catalog No. 03-338-2C


Add to cart

Description

Description

  • Conventional open-end drum thieves provide a quick method for sampling unmarked drums
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications

Specifications

0.84 oz., 25mL
0.75cm
Provides a quick method for sampling unmarked drums
109.2cm
0.84 oz.
Drum Sampler
0.29 in.
Yes
43 in.
Borosilicate Glass
25mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit