Learn More
DWK Life Sciences Kimble™ GL 45 Polypropylene Color Coded Screw Thread Caps
GREENER_CHOICESDP
Catalog No. 02542357 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Gray
Green
Pink
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-542-357 Green
02-542-356 Gray
02-542-358 Pink
02-542-5A White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-542-357 Supplier DWK Life Sciences Supplier No. 14395C457
Only null left
Add to Cart
Add to Cart
Autoclavable screw-thread caps provide secure closure for storage bottles and flasks.
  • Solid top polypropylene caps with internal molded seal rings
  • Caps are designed to fit GL 45 threaded Media Bottles and Flasks
  • Autoclavable 140°C
  • Colors available: Gray, Green, Pink, Blue, White, Orange
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Includes Internal molded seal rings
Material Polypropylene
Max. Temperature (English) 140°C
Shape Round
Product Type Screw Thread Cap
Closure Material Polypropylene
Closure Type Solid
Autoclavable Autoclavable