Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ GL 45 Polypropylene Color Coded Screw Thread Caps
GREENER_CHOICESDP

Catalog No. 025425A Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Gray
Green
Pink
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-542-5A White
02-542-356 Gray
02-542-357 Green
02-542-358 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-542-5A Supplier DWK Life Sciences Supplier No. 14395C451
Only null left
Add to Cart
Add to Cart

Autoclavable screw-thread caps provide secure closure for storage bottles and flasks.

  • Solid top polypropylene caps with internal molded seal rings
  • Caps are designed to fit GL 45 threaded Media Bottles and Flasks
  • Autoclavable 140°C
  • Colors available: Gray, Green, Pink, Blue, White, Orange
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 10 Cs.
Thread Size (Metric) GL 45
For Use With (Equipment) Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.