Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Glass Screw-Thread Sample Vials with PTFE Closure

Cylindrical shape provides solid balance. Flat bottom facilitates free-standing capability.

Manufacturer:  DWK Life Sciences 60940A12

 View more versions of this product

Catalog No. 03-340-60C


  Check Availability
Add to cart

Description

Description

Cylindrical shape provides solid balance. Flat bottom facilitates free-standing capability.

  • Vial top has a screw thread finish
  • Supplied with attached foam-backed, PTFE-lined urea closure

Specifications

Specifications

ASTM Type I, Class A
12.0mL
19mm
Borosilicate Glass
12mL
PTFE
3.2 dr.
Closed screw top
A
Clear
Sample Vial
67mm
Screw
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit