Learn More
DWK Life Sciences Kimble™ Glass Screw-Thread Sample Vials without Closure
Cylindrical shape provides solid balance. Flat bottom facilitates free-standing capability.
Manufacturer:  DWK Life Sciences 608102170
 View more versions of this product
Catalog No. 03-340-5R
Add to cart
Description
Description
  • Glass sample vial has a screw thread finish
  • KG-33 borosilicate glass
  • Ungraduated with no closure included
Specifications
Specifications
16.0 mL
18-400
70mm
Screw
1 dr.
21 mm
ASTM Type I, Class A
Screw
18-400
Sample Vial
Clear
Borosilicate Glass