Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimble™ Glass Screw-Thread Sample Vials without Closure

Cylindrical shape provides solid balance. Flat bottom facilitates free-standing capability.

Manufacturer:  Kimble™ 608101965

 View more versions of this product

Catalog No. 03-340-5S



  Check Availability
Add to cart

Specifications

Description

  • Glass sample vial has a screw thread finish
  • KG-33 borosilicate glass
  • Ungraduated with no closure included

Description & Specifications

Specifications

ASTM Type I, Class A
12.0mL
Clear
Sample Vial
65mm
Screw
Screw
19mm
Borosilicate Glass
3mL
15-425
0.8 dr.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit