Learn More
DWK Life Sciences Kimble™ Glass Screw-Thread Sample Vials without Closure Available on GSA/VA Contract for Federal Government customers only.
Cylindrical shape provides solid balance. Flat bottom facilitates free-standing capability.
Manufacturer:  DWK Life Sciences 608151760
 View more versions of this product
Catalog No. 03-339-17B
Add to cart
Description
Description
  • Glass sample vial has a screw thread finish
  • KG-33 borosilicate glass
  • Ungraduated with no closure included
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
USP Type I
Screw
Screw
2.16 dr.
17 mm
8.0 mL
15-425
60mm
Sample Vial
Amber
Borosilicate Glass