Learn More
DWK Life Sciences Kimble™ KIMAX™ Brand Filtering Flasks
Designed to afford maximum strength
Supplier: DWK Life Sciences 270502000
Description
- Heavy walled
- Necks tooled for uniform stopper fit with a thick flat flange
- Without side tabulation
- Durable, white ceramic enamel scale to indicate approximate volume
Certifications
ASTM Specification E1406, Type I requirements
Specifications
ASTM, E1406, Type I | |
2000 mL | |
1000 to 4000 mL | |
11.8 in. | |
16.8 cm |
6.6 in. | |
Borosilicate Glass | |
100 mL | |
30 cm | |
9 |