Learn More
DWK Life Sciences Kimble™ KIMAX™ Brand Filtering Flasks
Designed to afford maximum strength
$121.00 - $293.00
Specifications
Certifications/Compliance ASTM, E1406, Type I
Material Borosilicate Glass
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
10-310-88C
Accessories View Documents Promotion Details
DWK Life Sciences
270502000
2000 mL
Case for $217.00
Add to cart
10-310-88D
View Documents Promotion Details
DWK Life Sciences
270504000
4 L
Case for $293.00
Add to cart
10-310-88B
Accessories View Documents Promotion Details
DWK Life Sciences
270501000
1000 mL
Case for $121.00
Add to cart
10-310-88
Accessories View Documents
DWK Life Sciences
27050500
500 mL This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Heavy walled
  • Necks tooled for uniform stopper fit with a thick flat flange
  • Without side tabulation
  • Durable, white ceramic enamel scale to indicate approximate volume
Certifications
ASTM Specification E1406, Type I requirements
Specifications
Specifications
ASTM, E1406, Type I
Borosilicate Glass