Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ KIMAX™ KimCote™ Heavy Wall, Round Bottom Flask
The Kimble™ KIMAX™ KimCote™ Heavy Wall, Round Bottom Flask is a plastic-coated single neck flask useful as a rotary evaporator condensate collection flask or as a replacement for European designation KS 20/35.
Supplier: DWK Life Sciences KC25330500
This item is not returnable.
View return policy
Description
- Spherical socket joint
- Joint Size: 35/20 mm
- Autoclavable KimCote safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
500 mL | |
Spherical | |
Autoclavable | |
No | |
Round | |
Plastic Coated |
Borosilicate Glass (3.3) | |
35/25 | |
Reusable | |
Non-sterile | |
102 mm |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction