Learn More
DWK Life Sciences Kimble™ KIMAX™ KimCote™ Heavy Wall, Round Bottom Flask
The Kimble™ KIMAX™ KimCote™ Heavy Wall, Round Bottom Flask is a plastic-coated single neck flask useful as a rotary evaporator condensate collection flask or as a replacement for European designation KS 20/35.
Supplier:  DWK Life Sciences KC253302000
 View more versions of this product
Catalog No. 01-183-003
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Spherical socket joint
  • Joint Size: 35/20 mm
  • Autoclavable KimCote safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
2000 mL
Spherical
Autoclavable
No
Round
Plastic Coated
Borosilicate Glass 3.3
35/25
No
Non-sterile
161 mm